Learn More
IL-2 Mouse anti-Human, Brilliant Violet 650, Clone: 5344.111, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563467
Description
The 5344.111 antibody specifically binds to the multifunctional cytokine, human interleukin-2 (IL-2). IL-2 is produced by activated T cells and has multiple functions that can affect the growth, proliferation and differentiation of many different target cell types including T cells, B cells, NK cells, monocytes and macrophages. The immunogen used to generate the 5344.111 hybridoma was affinity-purified, recombinant human IL-2 protein.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will bespillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| IL-2 | |
| Monoclonal | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IL-2 protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 5344.111 | |
| IL2; Interleukin-2; T-cell growth factor; TCGF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.