Learn More
IL-1ra Mouse anti-Human, PE, Clone: AS17, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340525
Description
Anti-Hu–IL-1ra, clone AS17, is derived from the fusion of P3X63.Ag8.653 myeloma cells with splenocytes from BALB/c mice immunized with recombinant human IL-1 receptor antagonist.
Anti-Human Interleukin-1ra (Anti-Hu–IL-1ra) recognizes a 17- to 25-kilodalton (kd) polypeptide.
Specifications
| IL-1ra | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry | |
| AS17 | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Recombinant human IL-1 receptor antagonist | |
| 50 Tests | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.