missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1b Mouse anti-Human, PE, Clone: AS10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340516
This item is not returnable.
View return policy
Description
Anti-Hu–IL-1β, clone AS10, is derived from fusion of P3X63Ag8 myeloma cells with splenocytes from BALB/c mice immunized with recombinant human IL-1β.
Anti-Human Interleukin-1β (Anti-Hu–Il-1β) recognizes a 13- to 17-kilodalton (kd) polypeptide.
Specifications
| IL-1b | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry | |
| AS10 | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human IL-1β | |
| 50 Tests | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction