Learn More
IL-17A Rat anti-Mouse, Alexa Fluor 700, Clone: TC11-18H10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560820
Description
The TC11-18H10 monoclonal antibody specifically binds to recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCRαβ+CD4-CD8- thymocytes.
Specifications
| IL-17A | |
| Monoclonal | |
| 0.2mg/mL | |
| Interleukin-17A; Il17a; Cytotoxic T-lymphocyte-associated antigen; CTLA-8 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| TC11-18H10 | |
| Alexa Fluor 700 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Recombinant Mouse IL-17A Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.