missing translation for 'onlineSavingsMsg'
Learn More

IL-12 p70 Rat anti-Mouse, Unlabeled, Clone: 9A5, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554658

Product Code. 15827828

  • ONLINE EXCLUSIVE PRICE
    £377.00
    £361.35 / Each
    Save £15.65
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 9A5 antibody reacts with the p70 subunit of mouse IL-12. The immunogen used to generate the 9A5 hybrodoma was CHO-expressed recombinant mouse IL-12 p70. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 9A5
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: CHO-Expressed Recombinant Mouse IL-12 p70

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-12 p70
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CHO-Expressed Recombinant Mouse IL-12 p70
0.5 mg
Immunology
Mouse
IgG2b
ELISA
9A5
Unconjugated
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.