missing translation for 'onlineSavingsMsg'
Learn More

IL-12 p70 Rat anti-Human, Unlabeled, Clone: 20C2, BD

Product Code. 15857988
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15857988 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15857988 Supplier BD Biosciences Supplier No. 555065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The 20C2 antibody reacts with the biologically active form of human IL-12 p70 heterodimer, but not with the p40 monomer or homodimer. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p70
Applications ELISA
Classification Monoclonal
Clone 20C2
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.