Learn More
IL-12 (p40/p70) Mouse anti-Human, Unlabeled, Clone: C11.5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554573
Description
The C11.5 antibody reacts with human IL-12 p40 monomer and p70 heterodimer, but not p35 monomer. The immunogen used to generate the C11.5 hybridoma was the CHO-expressed recombinant human IL-12 p70 heterodimer. p40 has also been described as a subunit of IL-23 and thus it is possible that the C11.5 antibody will crossreact with IL-23.
Host Species: Mouse
Clone: C11.5
Isotype: IgG1
Species Reactivity: Human
Immunogen: CHO-expressed recombinant human IL-12 p70 heterodimer
Blocking Assay, Intracellular Staining
Specifications
| IL-12 (p40/p70) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| CHO-expressed recombinant human IL-12 p70 heterodimer | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 |
| Blocking Assay, Flow Cytometry | |
| C11.5 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.