missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Rat anti-Human and Viral, Unlabeled, Clone: JES3-9D7, BD
Rat Monoclonal Antibody
£170.00 - £311.86
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES3-9D7 |
| Concentration | 0.5mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15837738
|
BD Biosciences
554496 |
0.1 mg | |||||||
|
15847738
|
BD Biosciences
554497 |
0.5 mg |
£325.00 £311.86 / Each Save £13.14 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The JES3-9D7 antibody reacts with human IL-10 and viral IL-10. The immunogen used to generate the JES3-9D7 hybridoma was recombinant human IL-10 expressed in COS cells.
Specifications
| IL-10 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| CSIF; Cytokine synthesis inhibitory factor; TGIF |
| JES3-9D7 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Human, Virus | |
| COS-expressed Recombinant Human IL-10 | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title