Learn More
IL-1α Mouse anti-Human, PE, Clone: 364-3B3-14, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554561
Description
The 364-3B3-14 antibody reacts with human interleukin-1α (IL-1α). The immunogen used to generate the 364-3B3-14 hybridoma was recombinant human IL-1α. The 364-3B3-14 antibody does not cross-react with human IL-1β. This is a non-neutralizing antibody.
Host Species: Mouse
Clone: 364-3B3-14
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: recombinant human IL-1α
Intracellular Staining
Specifications
| IL-1α | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| recombinant human IL-1α | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 364-3B3-14 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.