Learn More
IL-1β Mouse anti-Human, Unlabeled, Clone: AS10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550007
Description
The AS10 antibody reacts with human interleukin-1β (IL-1β) which is also known as endogenous pyrogen (EP), leukocyte endogenous mediator (LEM), mononuclear cell factor (MCF) and lymphocyte-activating factor (LAF). IL-1β is a proinflammatory cytokine that is synthesized as a precursor of 31kDa and is converted intracellularly to the mature 17.5kDa form, after cleavage by the IL-1β-converting enzyme (ICE). In healthy individuals, IL-1β is secreted non-constitutively by blood monocytes, tissue macrophages and dendritic cells. IL-1β is also constitutively expressed in the human hypothalamus. Many malignant tumors express IL-1β as part of their neoplastic nature. The immunogen used to generate the AS10 hybridoma was recombinant human E. Coli-expressed recombinant IL-1β. The AS10 antibody does not cross react with mouse IL-1β. This is a neutralizing antibody.
Specifications
| IL-1β | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant human IL-1β | |
| 0.25 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Immunocytochemistry | |
| AS10 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.