Learn More
IKKγ Mouse, Unlabeled, Clone: 54, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611306
Description
In most cells, NF-κB is sequestered in an inactive cytoplasmic form via interactions with the inhibitory proteins IκBα, IκBβ, and IκBε. Cell
stimulation induces the release, activation, and nuclear translocation of NF-κB. Release of NF-κB results from the phosphorylation and subsequent proteolytic degradation of the IκB proteins. Two cytokine-inducible IκB kinases (IKKα and IKKβ) phosphorylate and target the IκB proteins for degradation by the ubiquitin pathway. These kinases are components of a 700-900kDa multisubunit complex that also contains NF-κB/RelA, IκBα, MEKK1, NIK, IKAP, and IKKγ/NEMO (NF-κB essential modulator). IKKγ contains two coiled-coil domains and a leucine zipper which allow it to form dimers and trimers that interact directly with IKKβ. IKKγ, essential for IKKα/IKKβ activation of NF-κB, is located functionally and physically upstream of these subunits. In addition, IKKγ enables the HTLV-1 Tax oncoprotein to interact with IKKβ, resulting in constitutive activation of NF-κB and maintenance of cellular transformation. Thus, IKKγ is an essential element of the IκB complex and an adaptor that mediates stable formation of Tax-IKK complexes.
Host Species: Mouse
Clone: 54
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse IKKγ/NEMO aa. 278-396
Formula Weight [Chemical]: 48kDa
Immunofluorescence, Western Blotting
Specifications
| IKKγ | |
| Monoclonal | |
| 250μg/mL | |
| NEMO | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 54 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse IKKγ/NEMO aa. 278-396 | |
| 50 μg | |
| Cell Biology | |
| Canine, Human, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.