missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Rat anti-Mouse, PE, Clone: R6-60.2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562033
This item is not returnable.
View return policy
Description
The R6-60.2 antibody monoclonal antibody specifically binds to mouse Immunoglobulin M (IgM) of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. The R6-60.2 antibody has not been shown to stimulate B-cell proliferation.
Specifications
| IgM | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Pooled Mouse Ig | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| R6-60.2 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction