missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2b, κ, Clone: MPC-11, BD

Product Code. 15829738
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829738 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829738 Supplier BD Biosciences Supplier No. 559530
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The MPC-11 antibody has unknown specificity. The MPC-11 cell line was adapted to continuous culture from the tranplantable Merwin Plasmacytoma-11, which was induced by intraperitoneal implantation of a Millipore diffusion chamber into a BALB/c mouse. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Functional Assay, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Blocking Assay, Functional Assay, Cytotoxicity Assay, Functional Assay
Clone MPC-11
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Content And Storage Undiluted at 4°C
Isotype IgG2b κ
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.