missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2b, κ, Alexa Fluor™ 488, Clone: 27-35, BD

Brand:  BD Biosciences 558716

Product Code. 15819608

  • ONLINE EXCLUSIVE PRICE
    £79.20
    £76.18 / Each
    Save £3.02
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-(dimethylamino) naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues.

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
Alexa Fluor 488
Affinity Purified
Undiluted at 4°C
27-35
Mouse
50 Tests
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.