missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Rat, FITC, Clone: R35-95, Isotype Control, BD

Brand:  BD Biosciences 553929

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10187143

  • £360.22 / Each

Estimated Shipment: 27-01-2022
Add to basket

Description

Description

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin

Flow Cytometry, Intracellular Staining, Isotype Control

Specifications

Specifications

Flow Cytometry, Western Blot
0.5mg/mL
Affinity Purified
Mouse Pooled Immunoglobulin
0.25mg
R35-95
FITC
Rat
IgG2a, k
Undiluted at 4°C
Videos
SDS
Documents
Certificates
Special Offers

Special Offers