Learn More
IgG2a Mouse anti-Rat, PE, Clone: RG7/1.30, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558067
Description
The Rg7/1.30 antibody reacts specifically with the Fc region of rat IgG2a. It does not react with other Ig isotypes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: RG7/1.30
Isotype: IgG2b, κ
Cross Reactivity: Rat
ELISA, Flow Cytometry
Specifications
| IgG2a | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat Pooled IgG | |
| 0.2 mg | |
| Immunology | |
| Rat | |
| IgG2b κ |
| Flow Cytometry | |
| RG7/1.30 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.