missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Mouse, APC, Clone: G155-178, Isotype Control, BD

Brand:  BD Biosciences 550882

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15866418

  • £541.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: TNP-keyhole limpet hemocyanin

Flow Cytometry, Isotype Control

Specifications

Specifications

Flow Cytometry
0.2mg/mL
Mouse
Affinity Purified
Non-human Primate
IgG2a κ
G155-178
APC
TNP-keyhole limpet hemocyanin
0.1 mg
Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers