Learn More
anti-IgG2a, κ, FITC, Clone: G155-178, Isotype control, BD
For low background screening on a variety of mouse and human tissues. anti-IgG2a, κ, FITC, Clone: G155-178, BD antibody is routinely tested by flow cytometric analysis.
Brand: BD Biosciences 555573
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Applications: Flow Cytometry, Isotype Control
Specifications
| Flow Cytometry | |
| G155-178 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| IgG2a κ |
| Monoclonal | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.