missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, κ, R-PE, Clone: R3-34, BD

Brand:  BD Biosciences 559318

Product Code. 15859698

  • ONLINE EXCLUSIVE PRICE
    £182.00
    £174.62 / Each
    Save £7.38
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
R-PE
Affinity Purified
Undiluted at 4°C
R3-34
Rat
100 Tests
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.