missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1 κ Mouse, PerCP-Cy5.5, Clone: MOPC-21, BD
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | IgG1 κ |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MOPC-21 |
| Concentration | 0.2mg/mL |
| Conjugate | PerCP-Cyanine5.5 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?