missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ Mouse, R-PE, Clone: MOPC-21, Isotype Control, BD
Description
- Host Species: Mouse
- Clone: MOPC-21
- Isotype: IgG1, κ
- Applications: Flow Cytometry, Isotype Control
- Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MOPC-21 |
| Conjugate | R-PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?