missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnetic Particles - DM Mouse anti-Mouse, BD IMag-DM, Clone: A85-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557983
This item is not returnable.
View return policy
Description
The A85-1 clone has been reported to react specifically with mouse IgG1 of Igh-Ca and Igh-Cb haplotypes, but reportedly does not react with other Ig isotypes.
- Host Species: Mouse
- Clone: A85-1
- Isotype: IgG1, κ
- Species Reactivity: Mouse
- Immunogen: Pooled Mouse IgG1
- Unlabeled
- Strain Host: LOU
Cell separation
Specifications
| Magnetic Particles - DM | |
| Monoclonal | |
| BD IMag-DM | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Pooled IgG1 | |
| 5 mL | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| A85-1 | |
| Ighg1; Immunoglobulin heavy constant gamma 1; Igh-4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction