missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, κ , Clone: R3-34, BD

Product Code. 15809678
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809678 0.25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809678 Supplier BD Biosciences Supplier No. 559072
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by immunocytochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Immunocytochemistry, Immunohistochemistry (Formalin), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed)
Clone R3-34
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Rat
Purification Method Affinity Purified
Quantity 0.25 mg
Content And Storage Undiluted at 4°C
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.