missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, λ, Clone: A110-1, BD

Product Code. 15899678
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15899678 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15899678 Supplier BD Biosciences Supplier No. 559157
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The A110-1 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, or rat cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone A110-1 was selected as an isotype control following screening for low background on a variety of mouse and rat tissues. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.

Blocking Assay, Co-Stimulation, Cytotoxicity Assay, ELISA, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone A110-1
Concentration 0.2mg/mL
Conjugate Violet Fluor 500 (V500)
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Target Species Human, Rat
Content And Storage Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.