Learn More
IgG Detector Mouse anti-Rabbit, PE, Clone: C101-359, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558553
Description
The PE Mouse anti-Rabbit IgG Detector is useful in verification of the conjugation of rabbit IgG isotype antibodies to the BD CBA Functional Beads. It does not react with rabbit IgM. It does not cross-react with mouse, rat, human or guinea pig immunoglobulins.
Flow Cytometry
Specifications
| IgG Detector | |
| Monoclonal | |
| PE | |
| Mouse | |
| 25 Tests | |
| Immunology | |
| Rabbit | |
| IgG1 κ |
| Flow Cytometry | |
| C101-359 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.