Learn More
IgG Mouse anti-Pig, Unlabeled, Clone: F007-1241, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 552554
Description
The F007-1241 antibody reacts with pig IgG, but not with pig IgM. The reactivity with individual pig IgG subclasses (IgG1, IgG2a, IgG2b, IgG3, IgG4) was not determined. It does not cross-react with mouse, rat, hamster, or rabbit immunoglobulins. The F007-1241 antibody does have some cross-reactivity with dog immunoglobulins.
Host Species: Mouse
Clone: F007-1241
Isotype: IgG1 κ
Species Reactivity: Porcine
Immunogen: Purified pig IgG
ELISA
Specifications
| IgG | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Purified pig IgG | |
| 0.5 mg | |
| Immunology | |
| Pig | |
| IgG1 κ |
| ELISA | |
| F007-1241 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.