missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Mouse anti-Human, APC-H7, Clone: G18-145, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561297
This item is not returnable.
View return policy
Description
The G18-145 monoclonal antibody specifically binds to the heavy chain of human immunoglobulin G subclasses: IgG1, IgG2, IgG3 and IgG4. The G18-145 antibody has been reported not to react with the heavy chains of other human immunoglobulin isotypes.
Specifications
| IgG | |
| Monoclonal | |
| APC-H7 | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G18-145 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction