Learn More
IgA Rat anti-Mouse, Unlabeled, Clone: C10-1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 556960
Description
The C10-1 antibody reacts specifically with mouse IgA of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with C10-1 mAb.
Host Species: Rat
Clone: C10-1
Isotype: IgG1, κ
Cross Reactivity: Mouse
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| IgA | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Pooled mouse IgA | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
| ELISA | |
| C10-1 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.