missing translation for 'onlineSavingsMsg'
Learn More
Learn More
interferon, alpha 16, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human IFNA16 protein.
Brand: Abnova H00003449-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MALSFSLLMAVLVLSYKSICSLGCDLPQTHSLGNRRALILLAQMGRISHFSCLKDRYDFGFPQEVFDGNQFQKAQAISAFHEMIQQTFNLFSTKDSSAAWDETLLDKFYIELFQQLNDLEACVTQEVGVEEIALMNEDSILAVRKYFQRITLYLMGKKYSPCAWEVVRAEIMRSFSFSTNLQKGLRRKDSpecifications
| interferon, alpha 16 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human IFNA16 protein. | |
| IFNA16 | |
| IFNA16 | |
| IFNA16 (NP_002164, 1 a.a. ∼ 189 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002173 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3449 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction