Learn More
IFN-γ Mouse anti-Human, NA/LE, Unlabeled, Clone: B27, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554698
Description
The B27 antibody reacts with human interferon-γ (IFN-γ). The immunogen used to generate the B27 hybridoma was E.coli-expressed recombinant human IFN-γ. This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. This antibody is routinely tested as a blocking control for intracellular staining. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| IFN-γ | |
| Monoclonal | |
| 1.0mg/mL | |
| IFNG; Interferon-gamma; Interferon-γ; Type II interferon; MAF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| B27 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Human E.coli-expressed IFN-γ | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.