missing translation for 'onlineSavingsMsg'
Learn More
Learn More
interferon induced transmembrane protein 3 (1-8U), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human IFITM3 protein.
Brand: Abnova H00010410-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MNHTVQTFFSPVNSGQPPNYEMLKEEHEVAVLGAPHNPAPPTSTVIHIRSETSVPDHVVWSLFNTLFMNPCCLGFIAFAYSVKSRDRKMVGDVTGAQAYASTAKCLNIWALILGILMTILLIVIPVLIFQAYGSpecifications
| interferon induced transmembrane protein 3 (1-8U) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human IFITM3 protein. | |
| IFITM3 | |
| 1-8U/IP15 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 10410 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC006794 | |
| IFITM3 | |
| IFITM3 (AAH06794, 1 a.a. ∼ 133 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction