Learn More
inducible T-cell co-stimulator, Mouse, Clone: 1G1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ICOS.
Brand: Abnova H00029851-M01.50ug
Description
The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation. [provided by RefSeq
Sequence: MKSGLWYFFLFCLRIKVLTGEINGSANYEMFIFHNGGVQILCKYPDIVQQFKMQLLKGGQILCDLTKTKGSGNTVSIKSLKFCHSQLSNNSVSFFLYNLDHSHANYYFCNLSIFDPPPFKVTLTGGYLHIYESQLCCQLKFWLPIGCAAFVVVCILGCILICWLTKKMSpecifications
| inducible T-cell co-stimulator | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC028006 | |
| ICOS | |
| ICOS (AAH28006, 1 a.a. ∼ 168 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 50 μg | |
| Yes | |
| 29851 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA | |
| 1G1 | |
| Mouse monoclonal antibody raised against a full-length recombinant ICOS. | |
| ICOS | |
| AILIM/CD278/MGC39850 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.