missing translation for 'onlineSavingsMsg'
Learn More

anti-ICOS/CD278 APC, Clone: ISA-3, Invitrogen™

ICOS/CD278 Monoclonal antibody A14773 from Invitrogen specifically detects ICOS/CD278 in Human samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A14773

Product Code. 13355614

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ICOS/CD278 Monoclonal antibody A14773 from Invitrogen specifically detects ICOS/CD278 in Human samples. It is validated for Flow Cytometry.

CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: ISA-3
Immunogen: Human ICOS
Formulation: 25 Tests Purified Antibody in proprietary buffer, with 0.09% sodium azide
Synonyms: AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Uniprot Accession: Q9Y6W8
Entrez GeneID: 29851
Gene Symbol: ICOS
This product detects ICOS/CD278 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Human ICOS.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICOS/CD278
Monoclonal
APC
Q9Y6W8
ICOS
Human ICOS
Primary
Human
Purified
Flow Cytometry
ISA-3
25 Tests Purified Antibody in proprietary buffer, with 0.09% sodium azide
AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Mouse
25 Tests
29851
Store at 4° C
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.