Learn More
anti-Iba1, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB5076-100UG
Description
Host Species: Goat
Isotype: IgG
Cross Reactivity: Rat, Guinea pig, Human. Predicted to work with Mouse, Pig, Macaque Monkey.
Immunogen: Synthetic peptide: C-TGPPAKKAISELP, corresponding to amino acids 135-147 of Human Iba1
Applications:
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| Iba1 | |
| 0.5mg/mL | |
| Goat | |
| Affinity Purified | |
| Cell Biology, Immunology, Neuroscience | |
| Guinea Pig, Human, Monkey, Mouse, Pig, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide: C-TGPPAKKAISELP, corresponding to amino acids 135-147 of Human Iba1 | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.