missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The STRO-1 antibody from Proteintech is a mouse monoclonal antibody generated with human cd34 positive bone marrow cells of human STRO-1. This antibody recognizes human antigen. The STRO-1 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
| Antigen | STRO-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | STRO-1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human CD34 positive bone marrow cells |
| Purification Method | Purified by precipitation and chromatography |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?