missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human HLA-DR (L243), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65218-1-IG-100UG
This item is not returnable.
View return policy
Description
The HLA-DR antibody from Proteintech is a mouse monoclonal antibody generated with human lymphoblastoid b-cell line rpmi 8866 of human HLA-DR. This antibody recognizes human antigen. The HLA-DR antibody has been validated for the following applications: FC analysis.Specifications
| HLA-DR | |
| Monoclonal | |
| 500 μg/mL | |
| HLA-DRA1 | |
| Human lymphoblastoid B-cell line RPMI 8866 | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| L243 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 3122 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction