missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD7 (4H9), Proteintech
Mouse Monoclonal Antibody
£47.00 - £145.00
Specifications
| Antigen | CD7 |
|---|---|
| Clone | 4H9 |
| Concentration | 500 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236664
|
Proteintech
65203-1-IG-100UG |
100 μg |
£50.00 £47.00 / 100µg Save £3.00 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235153
|
Proteintech
65203-1-IG-500UG |
500 μg |
£154.00 £145.00 / 500µg Save £9.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD7 antibody from Proteintech is a mouse monoclonal antibody generated with t-acute lymphoblastic leukemia cells of human CD7. This antibody recognizes human antigen. The CD7 antibody has been validated for the following applications: FC analysis.Specifications
| CD7 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 924 | |
| T-acute lymphoblastic leukemia cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| 4H9 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| GP40; LEU-9; TP41; Tp40 | |
| IgG2a κ | |
| Protein A purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title