Learn More
Anti-Human CD44 (F10-44-2), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65063-1-IG-500UG
 View more versions of this product
Product Code. 30237440
  • ONLINE EXCLUSIVE PRICE
    £154.00
    £145.00 / 500µg
    Save £9.00
    5% Off
Description
The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD44. This antibody recognizes human antigen. The CD44 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD44
Monoclonal
0.5 mg/mL
CDW44; CSPG8; ECMR-III; HCELL; IN; LHR; MC56; MDU2; MDU3; MGC10468; MIC4; MUTCH-I; Pgp1
Purified T cells from human lymph nodes
500 μg
Primary
Human
Flow Antibody
IgG2a κ
Flow Cytometry
F10-44-2
Unconjugated
Mouse
Affinity purification
RUO
960
Store at 2-8°C. Stable for one year after shipment.