missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD44 (F10-44-2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65063-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD44. This antibody recognizes human antigen. The CD44 antibody has been validated for the following applications: FC analysis.Specifications
| CD44 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CDW44; CSPG8; ECMR-III; HCELL; IN; LHR; MC56; MDU2; MDU3; MGC10468; MIC4; MUTCH-I; Pgp1 | |
| Purified T cells from human lymph nodes | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| F10-44-2 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 960 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction