Learn More
Anti-Human CD4 (OKT4), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65134-1-IG-500UG
Product Code. 30234795
  • ONLINE EXCLUSIVE PRICE
    £54.90
    £54.00 / 500µg
    Save £0.90
    1% Off
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral t cells of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD4
Monoclonal
0.5 mg/mL
CD4mut
Human peripheral T cells
500 μg
Primary
Human
Flow Antibody
IgG2b κ
Flow Cytometry
OKT4
Unconjugated
Mouse
Affinity purification
RUO
920
Store at 2-8°C. Stable for one year after shipment.