missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD20 (2H7), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65085-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with human tonsillar b cells of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC analysis.Specifications
| CD20 | |
| Monoclonal | |
| 500 μg/mL | |
| B1; Bp35; CD20; LEU-16; MGC3969; MS4A2; S7 | |
| Human tonsillar B cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2b κ |
| Flow Cytometry | |
| 2H7 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 931 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction