Learn More
Anti-Human CD20 (2H7), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65085-1-IG-100UG
 View more versions of this product
Product Code. 30235617
  • ONLINE EXCLUSIVE PRICE
    £35.00
    £33.00 / 100µg
    Save £2.00
    5% Off
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with human tonsillar b cells of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD20
Monoclonal
500 μg/mL
B1; Bp35; CD20; LEU-16; MGC3969; MS4A2; S7
Human tonsillar B cells
100 μg
Primary
Human
Flow Antibody
IgG2b κ
Flow Cytometry
2H7
Unconjugated
Mouse
Affinity purification
RUO
931
Store at 2-8°C. Stable for one year after shipment.