missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD14 (26IC), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65246-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC, ELISA analysis.Specifications
| CD14 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2b |
| Flow Cytometry, ELISA | |
| 26IC | |
| Unconjugated | |
| Affinity purification | |
| RUO | |
| 929 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction