missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD107a / LAMP1 (H4A3), Proteintech
Mouse Monoclonal Antibody
£47.00 - £140.00
Specifications
| Antigen | CD107a / LAMP1 |
|---|---|
| Clone | H4A3 |
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238028
|
Proteintech
65051-1-IG-500UG |
500 μg |
£157.00 £140.00 / 500µg Save £17.00 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235113
|
Proteintech
65051-1-IG-100UG |
100 μg |
£51.00 £47.00 / 100µg Save £4.00 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD107a / LAMP1 antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107a / LAMP1. This antibody recognizes human antigen. The CD107a / LAMP1 antibody has been validated for the following applications: WB, IF/ICC, FC (Intra) analysis.Specifications
| CD107a / LAMP1 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3916 | |
| Human adherent peripheral blood cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| H4A3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD107a; LAMPA; LGP120 | |
| IgG1 κ | |
| Protein G purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title