missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2C Polyclonal Antibody

HTR2C Polyclonal Antibody
Brand: Invitrogen PA533308
This item is not returnable.
View return policy
Description
HTR2C Polyclonal Antibody for IHC (P)
Specifications
| HTR2C | |
| Polyclonal | |
| Unconjugated | |
| HTR2C | |
| 5ht1c; 5-HT1C; 5-HT-1C; 5HT-1C; 5-HT1c receptor; 5-HT2C; 5-HT-2C; 5HT2C Receptor; 5-HT2C receptor; 5-HT2cR; 5HTR2C; 5-HTR2C; 5-hydroxytryptamine (serotonin) receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HGNC:5295; HTR1C; Htr2c; RP11-810O3.1; RP23-15B7.1; serotonin 1c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; serotonin receptor 2C; SR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3358 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28335 | |
| HTR2C | |
| Synthetic 16 amino acid peptide from C-terminus of human 5HT2C Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction