missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSV Type 1 gD Monoclonal Antibody (SKM/56)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518039
This item is not returnable.
View return policy
Description
.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.
Specifications
| HSV Type 1 gD | |
| Monoclonal | |
| 1.65 mg/mL | |
| PBS with 10mM sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA | |
| SKM/56 | |
| Unconjugated | |
| Herpes Simplex 1; Herpes Simplex I; HHV; HSV; HSV Type 1; HSV1 | |
| HSV-1. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction