Learn More
Hsp75/TRAP1 Mouse, Unlabeled, Clone: 42, BD
Mouse Monoclonal Antibody
£163.84 - £401.97
Specifications
| Antigen | Hsp75/TRAP1 |
|---|---|
| Clone | 42 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15866169
|
BD Biosciences
612344 |
50 μg |
£171.00 £163.84 / Each Save £7.16 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15876169
|
BD Biosciences
612345 |
150 μg |
£419.00 £401.97 / Each Save £17.03 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The hsp90 family of molecular chaperones is involved in a number of signal transduction pathways, including cell division, protein folding and transport. A new member of this family was recently identified as TRAP1 (Tumor necrosis factor receptor-associated protein) or hsp75. TRAP1 is a mitochondrial protein that is an ATPase and ATP-binding protein, rendering it similar to hsp90. There is a high degree of homology between TRAP1 and hsp90 in 6 domains, however one major difference is that a highly charged domain present in hsp90 is absent in TRAP1. TRAP1 has been shown to interact with the intracellular domain of the type 1 receptor for tumor necrosis factor (TNFR-1IC) and to the retinoblastoma protein (Rb). Signaling through TNFR-1 induces antiviral responses, fibroblast proliferation, cytotoxicity, etc. TRAP1 binds TNFR-1IC outside of the death domain leading to disruption in the signaling by TNFR-1. Rb has been shown to play a pivotal role in cellular differentiation and progression through the cell cycle. TRAP1 associates with Rb via an LxCxE motif, which is common among proteins that interact with Rb through SV40 T-antigen binding domain. Thus, TRAP1 is a new member of the hsp90 family that interacts with Rb and TNFR-1IC.
Immunofluorescence, Western Blotting
Specifications
| Hsp75/TRAP1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| TRAP1 |
| 42 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Human Hsp75/TRAP1 aa. 253-464 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.