missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydroxysteroid (17-beta) dehydrogenase 12 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HSD17B12.
Brand: Abnova H00051144-A01.50uL
This item is not returnable.
View return policy
Description
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.[supplied by OMIM
Sequence: SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNGSpecifications
| hydroxysteroid (17-beta) dehydrogenase 12 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant HSD17B12. | |
| HSD17B12 | |
| KAR/SDR12C1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 51144 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016142 | |
| HSD17B12 | |
| HSD17B12 (NP_057226, 203 a.a. ∼ 271 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto