Learn More
NDUFAF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522002
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Rhesus Monkey (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NDUFAF4 Polyclonal antibody specifically detects NDUFAF4 in Human samples. It is validated for Western Blot.Specifications
NDUFAF4 | |
Polyclonal | |
Unconjugated | |
NDUFAF4 | |
1110007M04Rik; 3000003G13Rik; AW214064; bA22L21.1; C6orf66; hormone-regulated proliferation associated protein 20; hormone-regulated proliferation-associated 20 kDa protein; hormone-regulated proliferation-associated protein of 20 kDa; Hormone-regulated proliferation-associated protein of 20 kDa homolog; hormone-regulated proliferation-associated protein, 20 kDa; Hrpap20; HSPC125; My013; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4; NADH dehydrogenase (ubiquinone) complex I, assembly factor 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4; NADH:ubiquinone oxidoreductase complex assembly factor 4; Ndufaf4; Protein HRPAP20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29078 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9P032 | |
NDUFAF4 | |
Full length human HRPAP20 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |