Learn More
HOXC8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527979
Description
PA5-27979 targets HOXC8 in WB applications and shows reactivity with Human samples. The PA5-27979 immunogen is recombit fragment corresponding to a region within amino acids 1 and 225 of Human HOXC8. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
Specifications
| HOXC8 | |
| Polyclonal | |
| Unconjugated | |
| P31273 | |
| HOXC8 | |
| Recombit fragment corresponding to a region within amino acids 1 and 225 of Human HOXC8 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| HOXC8 | |
| HOX3, HOX3A, Hox-3.1, mouse, homolog of, homeo box 3A, homeo box C8, homeobox protein Hox-3A, homeobox protein Hox-C8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3224 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.